missing translation for 'onlineSavingsMsg'
Learn More

2,4-Difluorobenzyl Bromide 98.0+%, TCI America™

Catalog No. B17105G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B17105G 5 g
B171025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B17105G Supplier TCI America Supplier No. B17105G

Chemical Identifiers

CAS 23915-07-3
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.018
MDL Number MFCD00011649
InChI Key IBLMYGXJKQIGSN-UHFFFAOYSA-N
Synonym 2,4-difluorobenzyl bromide, 1-bromomethyl-2,4-difluorobenzene, 2,4-difluorobenzylbromide, alpha-bromo-2,4-difluorotoluene, benzene, 1-bromomethyl-2,4-difluoro, 2,4-difluoro benzyl bromide, 1-bromomethyl-2,4-difluoro-benzene, 2-chloromethyl quinoline hcl salt, fr cf d1e, pubchem4911
PubChem CID 90297
IUPAC Name 1-(bromomethyl)-2,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)F)CBr

Specifications

Color Yellow
Quantity 5 g
UN Number 2920
Formula Weight 207.02
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,4-Difluorobenzyl Bromide