accessibility menu, dialog, popup

UserName

CAS RN 2386-60-9

IUPAC Name: butane-1-sulfonyl chloride
Synonyms: 1-butanesulfonyl chloride
Molecular Weight (g/mol): 156.62
Molecular Formula: C4H9ClO2S
InChi Key: WEDIIKBPDQQQJU-UHFFFAOYSA-N
SMILES: CCCCS(Cl)(=O)=O
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)