accessibility menu, dialog, popup

UserName

CAS RN 2386-53-0

IUPAC Name: sodium dodecane-1-sulfonate
Synonyms: dodecyl sulfonate, sodium salt
Molecular Weight (g/mol): 272.38
Molecular Formula: C12H25NaO3S
InChi Key: DAJSVUQLFFJUSX-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCCCCCS([O-])(=O)=O
Molecular Weight (g/mol) 
  • (9)
  • (5)
Quantity 
  • (3)
  • (2)
  • (6)
  • (3)
Percent Purity 
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
Physical Form 
  • (6)
  • (1)
  • (3)
Grade 
  • (2)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (9)
  • (9)
  • (5)
  • (3)
  • (2)
  • (6)
  • (3)
  • (4)
  • (1)
  • (3)
  • (3)
  • (2)
  • (6)
  • (1)
  • (3)
  • (2)
  • (1)
  • (3)