missing translation for 'onlineSavingsMsg'
Learn More

Dodecane-1-sulfonic acid sodium salt, LiChropur™ EMD-Millipore

Catalog No. M1183080025
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1183080025 25 g Glass
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1183080025 Supplier Technidata Supplier No. 1.18308.0025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ion pair reagents for analytical HPLC LiChropur

Chemical Identifiers

CAS 2386-53-0
Molecular Formula C12H25NaO3S
Molecular Weight (g/mol) 272.379
InChI Key DAJSVUQLFFJUSX-UHFFFAOYSA-M
Synonym sodium 1-dodecanesulfonate, sodium dodecane-1-sulfonate, 1-dodecanesulfonic acid sodium salt, sodium laurylsulfonate, 1-dodecanesulfonic acid, sodium salt, unii-4gy1kjw8si, sodium dodecyl sulfonate, 4gy1kjw8si, laurylsulfonic acid sodium salt, 1-dodecanesulfonic acid, sodium salt 1:1
PubChem CID 23665726
IUPAC Name sodium;dodecane-1-sulfonate
SMILES CCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

Density 330kg/m3
Packaging Glass
Quantity 25 g
Formula Weight 272.37
Grade Not Available
Chemical Name or Material Dodecane-1-sulfonic acid sodium salt
Recommended Storage Store at 5° to 30°C