CAS RN 237430-03-4
CAS RN 237430-03-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Alsterpaullone
A cell-permeable, potent, reversible, and ATP competitive inhibitor of GSK-3β (IC50 = 4 nM) and Cdk1/cyclin B (IC50 = 35 nM).