accessibility menu, dialog, popup

UserName

H3CONClNNH2OCH3

CAS RN 23680-84-4

IUPAC Name: 2-chloro-6,7-dimethoxyquinazolin-4-amine
Synonyms: 2-chloro-6,7-dimethoxyquinazolin-4-amine
Molecular Weight (g/mol): 239.66
Molecular Formula: C10H10ClN3O2
InChi Key: HWIIAAVGRHKSOJ-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(N)=NC(Cl)=NC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results