accessibility menu, dialog, popup

UserName

CAS RN 23672-07-3

CH3NNHOOCH3SH2NOO

CAS RN 23672-07-3

IUPAC Name: N-[(1-ethylpyrrolidin-2-yl)methyl]-2-methoxy-5-sulfamoylbenzamide
Synonyms: sulpiride sulpirida sulpiridum (+-)-sulpiride sulpyrid sulpirid
Molecular Weight (g/mol): 341.43
Molecular Formula: C15H23N3O4S
InChi Key: BGRJTUBHPOOWDU-UHFFFAOYNA-N
SMILES: CCN1CCCC1CNC(=O)C1=CC(=CC=C1OC)S(N)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results