missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-Sulpiride 98.0+%, TCI America™

Catalog No. I08215G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I08215G 5 g
I082125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I08215G Supplier TCI America Supplier No. I08215G
Only null left

Chemical Identifiers

CAS 23672-07-3
Molecular Formula C15H23N3O4S
Molecular Weight (g/mol) 341.426
MDL Number MFCD00055220
InChI Key BGRJTUBHPOOWDU-NSHDSACASA-N
Synonym levosulpiride, --sulpiride, s---sulpiride, levopraid, levopride, l-sulpiride, unii-jtg7r315lk, s-sulpiride, levosulpirida
PubChem CID 688272
ChEBI CHEBI:64119
IUPAC Name N-[[(2S)-1-ethylpyrrolidin-2-yl]methyl]-2-methoxy-5-sulfamoylbenzamide
SMILES CCN1CCCC1CNC(=O)C2=C(C=CC(=C2)S(=O)(=O)N)OC

Specifications

Melting Point 187°C
Color White-Yellow
Quantity 5 g
Formula Weight 341.43
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-Sulpiride
EINECSNumber (9)-2168
RTECSNumber BZ3400100
TSCA No
Recommended Storage Refrigerator