accessibility menu, dialog, popup

UserName

CAS RN 23611-75-8

IUPAC Name: methyl 6-chloropyrazine-2-carboxylate
Synonyms: 2-chloro-6-CO2ME-pyrazine
Molecular Weight (g/mol): 172.57
Molecular Formula: C6H5ClN2O2
InChi Key: MVVYUJFEXRODQA-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=CC(Cl)=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results