accessibility menu, dialog, popup

UserName

CAS RN 23602-63-3

CH3ClOOH

CAS RN 23602-63-3

IUPAC Name: 5-chloro-2-hydroxy-3-methylbenzaldehyde
Synonyms: 5-chloro-2-hydroxy-3-methylbenzaldehyde
Molecular Weight (g/mol): 170.59
Molecular Formula: C8H7ClO2
InChi Key: NSKZAOKQZDLHGO-UHFFFAOYSA-N
SMILES: CC1=C(O)C(C=O)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)