accessibility menu, dialog, popup

UserName

NH2

CAS RN 23357-46-2

IUPAC Name: 1,2,3,4-tetrahydronaphthalen-1-amine
Synonyms: 1,2,3,4-tetrahydronaphthalen-1-amine
Molecular Weight (g/mol): 147.22
Molecular Formula: C10H13N
InChi Key: JRZGPXSSNPTNMA-UHFFFAOYNA-N
SMILES: NC1CCCC2=CC=CC=C12
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results