accessibility menu, dialog, popup

UserName

H3CBrH2C

CAS RN 23074-36-4

IUPAC Name: 2-bromobut-1-ene
Synonyms: 1-butene, 2-bromo-
Molecular Weight (g/mol): 135
Molecular Formula: C4H7Br
InChi Key: HQMXRIGBXOFKIU-UHFFFAOYSA-N
SMILES: CCC(Br)=C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results