accessibility menu, dialog, popup

UserName

CAS RN 22978-25-2

IUPAC Name: 2-chloro-5-nitro-N-phenylbenzamide
Synonyms: 2-chloro-5-nitrobenzanilide
Molecular Weight (g/mol): 276.68
Molecular Formula: C13H9ClN2O3
InChi Key: DNTSIBUQMRRYIU-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(Cl)C(=C1)C(=O)NC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results