accessibility menu, dialog, popup

UserName

H3CN

CAS RN 2294-76-0

IUPAC Name: 2-pentylpyridine
Synonyms: 2-pentylpyridine
Molecular Weight (g/mol): 149.24
Molecular Formula: C10H15N
InChi Key: HSDXVAOHEOSTFZ-UHFFFAOYSA-N
SMILES: CCCCCC1=CC=CC=N1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)