missing translation for 'onlineSavingsMsg'
Learn More

2-Amylpyridine 98.0+%, TCI America™

Catalog No. A22785G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A22785G Supplier TCI America Supplier No. A22785G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2294-76-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00051828
InChI Key HSDXVAOHEOSTFZ-UHFFFAOYSA-N
Synonym 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8
PubChem CID 16800
IUPAC Name 2-pentylpyridine
SMILES CCCCCC1=CC=CC=N1

Specifications

Color Yellow
Boiling Point 207°C
Quantity 5 g
Formula Weight 149.24
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2-Amylpyridine