missing translation for 'onlineSavingsMsg'
Learn More

2-n-Pentylpyridine, 98%

Catalog No. AAB2212514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2212514 25 g
AAB2212506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2212514 Supplier Thermo Scientific Chemicals Supplier No. B2212514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2294-76-0 | C10H15N | 149.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2294-76-0
Molecular Formula C10H15N
Molecular Weight (g/mol) 149.237
MDL Number MFCD00051828
InChI Key HSDXVAOHEOSTFZ-UHFFFAOYSA-N
Synonym 2-amylpyridine, pyridine, 2-pentyl, 2-n-pentylpyridine, 2-n-amylpyridine, pyridine, pentyl, unii-9n74l1ud11, 2-amyl pyridine, 2-pentyl pyridine, 2-pentyl-pyridine, acmc-1cmx8
PubChem CID 16800
IUPAC Name 2-pentylpyridine
SMILES CCCCCC1=CC=CC=N1

Specifications

Density 0.903
Boiling Point 203°C to 204°C
Flash Point 79°C (174°F)
Odor Pleasant
Refractive Index 1.489
Quantity 25 g
Beilstein 2772
Formula Weight 149.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-n-Pentylpyridine
Show More Show Less
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement

H315-H319-H335-H227

Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 218-937-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only