accessibility menu, dialog, popup

UserName

CAS RN 22888-70-6

CH3OOHOOOHOOHOHOHO

CAS RN 22888-70-6

IUPAC Name: 3,5,7-trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-2,3-dihydro-1,4-benzodioxin-6-yl]-3,4-dihydro-2H-1-benzopyran-4-one
Synonyms: silymarin silybin silibin hepatos lagosa silybinin silliver 2,3-dehydrosilybin silybine silibinina
Molecular Weight (g/mol): 482.44
Molecular Formula: C25H22O10
InChi Key: SEBFKMXJBCUCAI-UHFFFAOYNA-N
SMILES: COC1=CC(=CC=C1O)C1OC2=CC(=CC=C2OC1CO)C1OC2=CC(O)=CC(O)=C2C(=O)C1O
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results