missing translation for 'onlineSavingsMsg'
Learn More

phyproof™ Silybin A, ≥95% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002238
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111002238 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111002238 Supplier Merck Emd Millipore Supplier No. PHL8265310MG

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Specifications

CAS 22888-70-6
Product Line phyproof
Linear Formula C25H22O10
MDL Number MFCD00872186
Type Reference Substance
Quantity 10 mg
Formula Weight 482.44
Percent Purity ≥95% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG