accessibility menu, dialog, popup

UserName

CAS RN 228121-39-9

IUPAC Name: λ¹-rhodium(1+) (1Z,5Z)-cycloocta-1,5-diene (2R,5R)-1-{2-[(2R,5R)-2,5-diethylphospholan-1-yl]phenyl}-2,5-diethylphospholane tetrafluoroboranuide
Synonyms: λ¹-rhodium(1+) (R,R)-ethyl-duphos 1,5-cyclooctadiene, (Z,Z)- tetrafluoroborate
Molecular Weight (g/mol): 660.37
Molecular Formula: C30H48BF4P2Rh
InChi Key: DSYBEQKPSKLNMC-RTXTXWNLNA-N
SMILES: [Rh+].F[B-](F)(F)F.C1C\C=C/CC\C=C/1.CC[C@@H]1CC[C@@H](CC)P1C1=C(C=CC=C1)P1[C@H](CC)CC[C@H]1CC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results