accessibility menu, dialog, popup

UserName

PPdPClPhPhPhPhPhPhPh

CAS RN 22784-59-4

IUPAC Name: benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphane)
Synonyms: benzyl(chloro)palladiumbis(ylium); bis(triphenylphosphine)
Molecular Weight (g/mol): 757.59
Molecular Formula: C43H37ClP2Pd
InChi Key: GYAXNTWMUGUEJY-UHFFFAOYSA-M
SMILES: Cl[Pd++]CC1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1
Physical Form 
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)