accessibility menu, dialog, popup

UserName

OHBrO

CAS RN 22532-62-3

IUPAC Name: 4-bromo-2-hydroxybenzaldehyde
Synonyms: 4-bromo-2-hydroxybenzaldehyde
Molecular Weight (g/mol): 201.02
Molecular Formula: C7H5BrO2
InChi Key: HXTWKHXDFATMSP-UHFFFAOYSA-N
SMILES: OC1=CC(Br)=CC=C1C=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)