accessibility menu, dialog, popup

UserName

CAS RN 22499-12-3

IUPAC Name: 2-(2-methylpropoxy)-1,2-diphenylethan-1-one
Synonyms: 2-(2-methylpropoxy)-1,2-diphenylethanone
Molecular Weight (g/mol): 268.36
Molecular Formula: C18H20O2
InChi Key: JMVZGKVGQDHWOI-UHFFFAOYNA-N
SMILES: CC(C)COC(C(=O)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results