accessibility menu, dialog, popup

UserName

CH2OO

CAS RN 22408-41-9

IUPAC Name: 2-(ethenyloxy)oxane
Synonyms: 2-(ethenyloxy)oxane
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: BYUNYALHUMSCSA-UHFFFAOYNA-N
SMILES: C=COC1CCCCO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results