accessibility menu, dialog, popup

UserName

II

CAS RN 2236-52-4

IUPAC Name: 2,2'-diiodo-1,1'-biphenyl
Synonyms: 2,2'-diiodo-1,1'-biphenyl
Molecular Weight (g/mol): 406.01
Molecular Formula: C12H8I2
InChi Key: OZVRXSGTNWILMN-UHFFFAOYSA-N
SMILES: IC1=CC=CC=C1C1=CC=CC=C1I
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results