missing translation for 'onlineSavingsMsg'
Learn More

2,2'-Diiodobiphenyl 98.0+%, TCI America™

Catalog No. D27451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D27451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D27451G Supplier TCI America Supplier No. D27451G
Only null left

Chemical Identifiers

CAS 2236-52-4
Molecular Formula C12H8I2
Molecular Weight (g/mol) 406.005
MDL Number MFCD00017622
InChI Key OZVRXSGTNWILMN-UHFFFAOYSA-N
PubChem CID 137510
IUPAC Name 1-iodo-2-(2-iodophenyl)benzene
SMILES C1=CC=C(C(=C1)C2=CC=CC=C2I)I

Specifications

Melting Point 110°C
Color White-Yellow
Quantity 1 g
UN Number 3152
Formula Weight 406.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,2′-Diiodobiphenyl