CAS RN 2236-01-3
IUPAC Name:
(2-oxopropyl)triphenylphosphanium bromide
Synonyms:
(2-oxopropyl)triphenylphosphanium bromide
Molecular Weight (g/mol):
399.27
Molecular Formula:
C21H20BrOP
InChi Key:
KZBWHCFTAAHIJJ-UHFFFAOYSA-M
SMILES:
[Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Acetonyltriphenylphosphonium bromide, 98+%
Acetonyltriphenylphosphonium bromide, 98+%
CAS: 2236-01-3 Molecular Formula: C21H20BrOP Molecular Weight (g/mol): 399.27 MDL Number: MFCD00051863 InChI Key: KZBWHCFTAAHIJJ-UHFFFAOYSA-M Synonym: acetonyltriphenylphosphonium bromide,acetonyl triphenylphosphonium bromide,2-oxopropyl triphenylphosphanium bromide,phosphonium, 2-oxopropyl triphenyl-, bromide,acetonyltriphenylphosphoniumbromide,acmc-1cmpe,2-oxopropyl-triphenylphosphonium bromide,2-oxopropyl triphenylphosphonium bromide,2-oxopropyl triphenyl phosphanium bromide,phosphonium, 2-oxopropyl triphenyl-,bromide 1:1 PubChem CID: 638158 IUPAC Name: 2-oxopropyl(triphenyl)phosphanium;bromide SMILES: [Br-].CC(=O)C[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1