accessibility menu, dialog, popup

UserName

H3CSO

CAS RN 222554-30-5

IUPAC Name: 4-hexylthiophene-2-carbaldehyde
Synonyms: 4-hexylthiophene-2-carbaldehyde
Molecular Weight (g/mol): 196.31
Molecular Formula: C11H16OS
InChi Key: XZSDWKHOEIKBDB-UHFFFAOYSA-N
SMILES: CCCCCCC1=CSC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results