CAS RN 22237-12-3
IUPAC Name:
(3-amino-4-methylphenyl)boronic acid hydrochloride
Synonyms:
3-amino-4-methylphenylboronic acid hydrochloride
Molecular Weight (g/mol):
187.43
Molecular Formula:
C7H11BClNO2
InChi Key:
BKFDOPYYSBQWIK-UHFFFAOYSA-N
SMILES:
Cl.CC1=CC=C(C=C1N)B(O)O
1
–
2
of
2
results
1
3-Amino-4-methylphenylboronic acid, 98%, Thermo Scientific™
CAS: 22237-12-3 Molecular Formula: C7H10BNO2 Molecular Weight (g/mol): 150.98 InChI Key: PLTGUDDQNWJILD-UHFFFAOYSA-N Synonym: 3-amino-4-methylphenyl boronic acid,3-amino-4-methylbenzeneboronic acid,3-amino-4-methyl-phenyl boronic acid,3-amino-p-tolylboronic acid,boronic acid, 3-amino-4-methylphenyl,acmc-20agxu,pubchem1745,3-amino-4-methylphenylboranic acid,3-amino4methyl phenyl boronic acid PubChem CID: 2737803 IUPAC Name: (3-amino-4-methylphenyl)boronic acid SMILES: B(C1=CC(=C(C=C1)C)N)(O)O