missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. A27451G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A27451G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A27451G Supplier TCI America Supplier No. A27451G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Chemical Name or Material 3-Amino-4-methylphenylboronic Acid (contains varying amounts of Anhydride)
CAS 22237-12-3
Molecular Formula C7H11BClNO2
MDL Number MFCD01632201
Quantity 1 g
Synonym 3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
InChI Key BKFDOPYYSBQWIK-UHFFFAOYSA-N
SMILES Cl.CC1=CC=C(C=C1N)B(O)O
IUPAC Name (3-amino-4-methylphenyl)boronic acid hydrochloride
Molecular Weight (g/mol) 187.43
PubChem CID 2737803
Formula Weight 150.97
Physical Form Crystalline Powder
Show More Show Less