missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methylphenylboronic acid, 98%, Thermo Scientific™

Catalog No. p-7077533
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC455180010 1g
AC455180050 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC455180010 Supplier Thermo Scientific Chemicals Supplier No. 455180010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 22237-12-3
Molecular Formula C7H10BNO2
Molecular Weight (g/mol) 150.98
InChI Key PLTGUDDQNWJILD-UHFFFAOYSA-N
Synonym 3-amino-4-methylphenyl boronic acid, 3-amino-4-methylbenzeneboronic acid, 3-amino-4-methyl-phenyl boronic acid, 3-amino-p-tolylboronic acid, boronic acid, 3-amino-4-methylphenyl, acmc-20agxu, pubchem1745, 3-amino-4-methylphenylboranic acid, 3-amino4methyl phenyl boronic acid
PubChem CID 2737803
IUPAC Name (3-amino-4-methylphenyl)boronic acid
SMILES B(C1=CC(=C(C=C1)C)N)(O)O

Specifications

Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1g
Formula Weight 150.98
Percent Purity 98%
Chemical Name or Material 3-Amino-4-methylphenylboronic acid
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only