accessibility menu, dialog, popup

UserName

CAS RN 2215-77-2

IUPAC Name: 4-phenoxybenzoic acid
Synonyms: P-phenoxybenzoic acid
Molecular Weight (g/mol): 214.22
Molecular Formula: C13H10O3
InChi Key: RYAQFHLUEMJOMF-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C(OC2=CC=CC=C2)C=C1
Percent Purity 
  • (2)
  • (3)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)