accessibility menu, dialog, popup

UserName

CAS RN 22110-53-8

IUPAC Name: 1-isocyanoadamantane
Synonyms: 1-isocyanoadamantane
Molecular Weight (g/mol): 161.25
Molecular Formula: C11H15N
InChi Key: RPRJRJNIFAYHRF-UHFFFAOYSA-N
SMILES: [C-]#[N+]C12CC3CC(CC(C3)C1)C2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results