accessibility menu, dialog, popup

UserName

HOOBrHO

CAS RN 2208-15-3

IUPAC Name: 4-bromo-3-hydroxynaphthalene-2-carboxylic acid
Synonyms: 4-bromo-3-hydroxynaphthalene-2-carboxylic acid
Molecular Weight (g/mol): 267.08
Molecular Formula: C11H7BrO3
InChi Key: HBUFBIFPUAHIDX-UHFFFAOYSA-N
SMILES: OC(=O)C1=C(O)C(Br)=C2C=CC=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results