accessibility menu, dialog, popup

UserName

OHBHON

CAS RN 220616-39-7

IUPAC Name: [3-(cyanomethyl)phenyl]boronic acid
Synonyms: 3-(cyanomethyl)phenylboronic acid
Molecular Weight (g/mol): 160.97
Molecular Formula: C8H8BNO2
InChi Key: JFMYJQMAPRBDFF-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=CC(CC#N)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results