accessibility menu, dialog, popup

UserName

CAS RN 22029-36-3

IUPAC Name: ethane-1,2-diamine; sulfuric acid
Synonyms: ethylenediamine; sulfuric acid
Molecular Weight (g/mol): 158.17
Molecular Formula: C2H10N2O4S
InChi Key: BNZCDZDLTIHJAC-UHFFFAOYSA-N
SMILES: NCCN.OS(O)(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results