CAS RN 21725-69-9
IUPAC Name:
2,3-dihydro-1,2-benzoxazol-3-one
Synonyms:
2H-1,2-benzoxazol-3-one
Molecular Weight (g/mol):
135.12
Molecular Formula:
C7H5NO2
InChi Key:
QLDQYRDCPNBPII-UHFFFAOYSA-N
SMILES:
O=C1NOC2=CC=CC=C12
1
–
1
of
1
results
1
Benzo[d]isoxazol-3-ol, 96%, Thermo Scientific™
CAS: 21725-69-9 Molecular Formula: C7H5NO2 Molecular Weight (g/mol): 135.12 InChI Key: QLDQYRDCPNBPII-UHFFFAOYSA-N Synonym: benzo d isoxazol-3-ol,1,2-benzisoxazol-3 2h-one,1,2-benzoxazol-3-ol,1,2-benzisoxazol-3-ol,3-hydroxybenzisoxazole,3-hydroxy-1,2-benzisoxazole,1,2-benzisoxazolin-3-one,1,2-benzisoxazol-3-ol 8ci,2h-1,2-benzoxazol-3-one,benzisoxazolone PubChem CID: 210830 IUPAC Name: 1,2-benzoxazol-3-one SMILES: C1=CC=C2C(=C1)C(=O)NO2