CAS RN 2169-87-1
CAS RN 2169-87-1
IUPAC Name:
naphthalene-2,3-dicarboxylic acid
Synonyms:
naphthalene-2,3-dicarboxylic acid
Molecular Weight (g/mol):
216.19
Molecular Formula:
C12H8O4
InChi Key:
KHARCSTZAGNHOT-UHFFFAOYSA-N
SMILES:
OC(=O)C1=C(C=C2C=CC=CC2=C1)C(O)=O
1
–
1
of
1
results
1
2,3-Naphthalenedicarboxylic Acid 98.0+%, TCI America™
CAS: 2169-87-1 Molecular Formula: C12H8O4 Molecular Weight (g/mol): 216.192 MDL Number: MFCD00004104 InChI Key: KHARCSTZAGNHOT-UHFFFAOYSA-N Synonym: 2,3-naphthalenedicarboxylic acid,2,3-naphthalenedicarboxylicacid,acmc-1cb2t,ghl.pd_mitscher_leg0.770 PubChem CID: 95073 IUPAC Name: naphthalene-2,3-dicarboxylic acid SMILES: C1=CC=C2C=C(C(=CC2=C1)C(=O)O)C(=O)O