missing translation for 'onlineSavingsMsg'
Learn More

2,3-Naphthalenedicarboxylic Acid 98.0+%, TCI America™

$225.56 - $682.24

Chemical Identifiers

CAS 2169-87-1
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.192
MDL Number MFCD00004104
InChI Key KHARCSTZAGNHOT-UHFFFAOYSA-N
Synonym 2,3-naphthalenedicarboxylic acid, 2,3-naphthalenedicarboxylicacid, acmc-1cb2t, ghl.pd_mitscher_leg0.770
PubChem CID 95073
IUPAC Name naphthalene-2,3-dicarboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)C(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N05265G
View Documents
TCI America
N05265G
5 g N/A
Only null left
 
N052625G
View Documents
TCI America
N052625G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

2169-87-1
216.192
KHARCSTZAGNHOT-UHFFFAOYSA-N
95073
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)C(=O)O
C12H8O4
MFCD00004104
2,3-naphthalenedicarboxylic acid, 2,3-naphthalenedicarboxylicacid, acmc-1cb2t, ghl.pd_mitscher_leg0.770
naphthalene-2,3-dicarboxylic acid

Specifications

2169-87-1
White
MFCD00004104
2,3-naphthalenedicarboxylic acid, 2,3-naphthalenedicarboxylicacid, acmc-1cb2t, ghl.pd_mitscher_leg0.770
C1=CC=C2C=C(C(=CC2=C1)C(=O)O)C(=O)O
216.192
216.19
Crystalline Powder
246°C
C12H8O4
5 g
KHARCSTZAGNHOT-UHFFFAOYSA-N
naphthalene-2,3-dicarboxylic acid
95073
≥98.0% (GC,T)
2,3-Naphthalenedicarboxylic Acid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'