accessibility menu, dialog, popup

UserName

CAS RN 21663-61-6

IUPAC Name: 2,5-dichlorobenzonitrile
Synonyms: 2,5-dichlorobenzonitrile
Molecular Weight (g/mol): 172.01
Molecular Formula: C7H3Cl2N
InChi Key: LNGWRTKJZCBXGT-UHFFFAOYSA-N
SMILES: ClC1=CC=C(Cl)C(=C1)C#N
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)