accessibility menu, dialog, popup

UserName

CH3OOOHHO

CAS RN 2150-46-1

IUPAC Name: methyl 2,5-dihydroxybenzoate
Synonyms: methyl dihydroxybenzoate
Molecular Weight (g/mol): 168.15
Molecular Formula: C8H8O4
InChi Key: XGDPKUKRQHHZTH-UHFFFAOYSA-N
SMILES: COC(=O)C1=CC(O)=CC=C1O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)