accessibility menu, dialog, popup

UserName

CAS RN 21490-63-1

IUPAC Name: (2S,3S)-2,3-dimethyloxirane
Synonyms: (2S,3S)-2,3-dimethyloxirane
Molecular Weight (g/mol): 72.11
Molecular Formula: C4H8O
InChi Key: PQXKWPLDPFFDJP-IMJSIDKUSA-N
SMILES: C[C@@H]1O[C@H]1C
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)