accessibility menu, dialog, popup

UserName

H3COOCH3

CAS RN 21470-41-7

IUPAC Name: 4,4'-bis(pentyloxy)-1,1'-biphenyl
Synonyms: 4,4'-bis(pentyloxy)-1,1'-biphenyl
Molecular Weight (g/mol): 326.48
Molecular Formula: C22H30O2
InChi Key: XTKHESQIKTWMCD-UHFFFAOYSA-N
SMILES: CCCCCOC1=CC=C(C=C1)C1=CC=C(OCCCCC)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results