accessibility menu, dialog, popup

UserName

H3COOCH2

CAS RN 2146-71-6

IUPAC Name: ethenyl dodecanoate
Synonyms: ethenyl dodecanoate
Molecular Weight (g/mol): 226.36
Molecular Formula: C14H26O2
InChi Key: GLVVKKSPKXTQRB-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCC(=O)OC=C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results