accessibility menu, dialog, popup

UserName

CAS RN 2122-63-6

NH2IOHO

CAS RN 2122-63-6

IUPAC Name: 4-amino-3-iodobenzoic acid
Synonyms: benzoic acid, 4-amino-3-iodo-
Molecular Weight (g/mol): 263.03
Molecular Formula: C7H6INO2
InChi Key: FPLDDZRJTOXFCB-UHFFFAOYSA-N
SMILES: NC1=CC=C(C=C1I)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results