accessibility menu, dialog, popup

UserName

NOOH2NFF

CAS RN 211693-73-1

IUPAC Name: 2,3-difluoro-6-nitroaniline
Synonyms: 2,3-difluoro-6-nitroaniline
Molecular Weight (g/mol): 174.11
Molecular Formula: C6H4F2N2O2
InChi Key: DIPGYZSCGXBTEU-UHFFFAOYSA-N
SMILES: NC1=C(C=CC(F)=C1F)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results