accessibility menu, dialog, popup

UserName

NH2NH2NNH2S

CAS RN 2114-02-5

IUPAC Name: (diaminomethylidene)thiourea
Synonyms: diaminomethylidenethiourea
Molecular Weight (g/mol): 118.16
Molecular Formula: C2H6N4S
InChi Key: OKGXJRGLYVRVNE-UHFFFAOYSA-N
SMILES: NC(N)=NC(N)=S
Percent Purity 
  • (3)
Physical Form 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results