accessibility menu, dialog, popup

UserName

OHBrF

CAS RN 2105-94-4

IUPAC Name: 4-bromo-2-fluorophenol
Synonyms: 4-bromo-2-fluorophenol
Molecular Weight (g/mol): 191
Molecular Formula: C6H4BrFO
InChi Key: RYVOZMPTISNBDB-UHFFFAOYSA-N
SMILES: OC1=CC=C(Br)C=C1F
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)