missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-fluorophenol 98.0+%, TCI America™

Catalog No. B18355G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18355G 5 g
B183525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B18355G Supplier TCI America Supplier No. B18355G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2105-94-4
Molecular Formula C6H4BrFO
Molecular Weight (g/mol) 191.00
MDL Number MFCD00011722
InChI Key RYVOZMPTISNBDB-UHFFFAOYSA-N
Synonym 2-fluoro-4-bromophenol, 4-bromo-2-fluoro-phenol, phenol, 4-bromo-2-fluoro, 4-bromo-2-fluoro phenol, pubchem1488, p-bromo-o-fluorophenol, 2-fluoro-4-bromo phenol, acmc-209fh4, ksc204a3n, 3-fluoro-4-hydroxy-bromobenzene
PubChem CID 2724981
IUPAC Name 4-bromo-2-fluorophenol
SMILES OC1=CC=C(Br)C=C1F

Specifications

Color Yellow
Boiling Point 90°C
Quantity 5 g
Formula Weight 191.00
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorophenol