accessibility menu, dialog, popup

UserName

CAS RN 2103-64-2

HOOOHOHOOOH

CAS RN 2103-64-2

IUPAC Name: 3',4',5',6'-tetrahydroxy-3H-spiro[2-benzofuran-1,9'-xanthen]-3-one
Synonyms: gallein
Molecular Weight (g/mol): 364.31
Molecular Formula: C20H12O7
InChi Key: PHLYOKFVXIVOJC-UHFFFAOYSA-N
SMILES: OC1=CC=C2C(OC3=C(O)C(O)=CC=C3C22OC(=O)C3=CC=CC=C23)=C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results