CAS RN 2100-25-6
CAS RN 2100-25-6
IUPAC Name:
3-iodo-1,2,4,5-tetramethylbenzene
Synonyms:
3-iodo-1,2,4,5-tetramethylbenzene
Molecular Weight (g/mol):
260.12
Molecular Formula:
C10H13I
InChi Key:
MCRSYYLHVQGFNR-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(C)C(I)=C1C
1
–
1
of
1
results
1
3-Iodo-1,2,4,5-tetramethylbenzene 98.0+%, TCI America™
CAS: 2100-25-6 Molecular Formula: C10H13I Molecular Weight (g/mol): 260.12 MDL Number: MFCD00013705 InChI Key: MCRSYYLHVQGFNR-UHFFFAOYSA-N Synonym: 2,3,5,6-tetramethyliodobenzene,iododurene,3-iododurene,benzene, 3-iodo-1,2,4,5-tetramethyl,1-iodo-2,3,5,6-tetramethylbenzene,3-iodo-1,2,4,5-tetramethyl-benzene,acmc-209fgl,maybridge1_002808,3-iodo-1,4,5-tetramethylbenzene,iodo-2,3,5,6-tetramethylbenzene PubChem CID: 75011 IUPAC Name: 3-iodo-1,2,4,5-tetramethylbenzene SMILES: CC1=CC(C)=C(C)C(I)=C1C